Tripotassium orthophosphate monohydrate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Potassium phosphate monohydrate, Potassium phosphate tribasic monohydrate, Potassium phosphate hydrate
139000-2KG
139000-500G
BT139000-2KGEA
339
USD
BT139000-2KG
BT139000-500G
Tripotassium orthophosphate monohydrate ≥95%
Tripotassium orthophosphate monohydrate
CAS: 27176-10-9; EC No: 231-907-1; MDL No: MFCD00150410
Crystalline/Granular; Linear Formula: K3PO4·H2O; MW: 230.28
Density (g/mL): 2.564
Hygroscopic
- Hygroscopic
- Granular
Formula:
K₃O₄P·1H₂O Density: 2.564 g/mL |
MDL Number:
MFCD00150410 CAS Number: 27176-10-9 EINECS: 231-907-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...