Perfluoroheptane (mixture of isomers) ≥98%

Supplier: BeanTown Chemical

Synonyms: Hexadecafluoroheptane

138985-100G 138985-25G
BT138985-100GEA 160 USD
BT138985-100G BT138985-25G
Perfluoroheptane (mixture of isomers) ≥98%
Perfluoroheptane (mixture of isomers)

CAS: 335-57-9; EC No: 206-392-1; MDL No: MFCD00040339; RTECS: MJ0875000 Liquid; Linear Formula: CF3(CF2)5CF3; Molecular Formula: C7F16; MW: 388.05 Boiling Point: 82-84° Density (g/mL): 1.745; Refractive Index: 1.265


  • RTECS: MJ0875000
  • Refractive index: 1.265
  • Liquid

Formula: C₇F₁₆
MW: 388.05 g/mol
Boiling Pt: 82…84 °C
Density: 1.745 g/mL
MDL Number: MFCD00040339
CAS Number: 335-57-9
EINECS: 206-392-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR