2-(Diphenylphosphino)biphenyl
Supplier: BeanTown Chemical
|
Warning
|
138975-25G
138975-5G
BT138975-25GEA
352
USD
BT138975-25G
BT138975-5G
2-(Diphenylphosphino)biphenyl
2-(Diphenylphosphino)biphenyl
CAS: 13885-09-1
Solid; Molecular Formula: C24H19P; MW: 338.38
- Solid
Formula:
C₂₄H₁₉P MW: 338.38 g/mol |
MDL Number:
MFCD11559063 CAS Number: 13885-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...