N-(1-Naphthyl)ethylenediamine dihydrochloride ≥96%

Supplier: BeanTown Chemical

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

138970-10G 138970-50G
BT138970-10GEA 60.34 USD
BT138970-10G BT138970-50G
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥96%
N-(1-Naphthyl)ethylenediamine dihydrochloride

CAS: 1465-25-4; EC No: 215-981-2; MDL No: MFCD00012556; RTECS: KV5330000 Powder; Linear Formula: C10H7NHCH2CH2NH2·2HCl; Molecular Formula: C12H14N2·2HCl; MW: 259.18 Melting Point: 194-198° (decomposes) Hygroscopic , Light Sensitive


  • RTECS: KV5330000
  • Hygroscopic, Light Sensitive
  • Powder

Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 194…198 °C (dec.)
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR