4-Amylcyclohexanol (cis- and trans- mixture) ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Pentylcyclohexanol

138960-25G 138960-5G
BT138960-25GEA 277 USD
BT138960-25G BT138960-5G
4-Amylcyclohexanol (cis- and trans- mixture) ≥98%
4-Amylcyclohexanol (cis- and trans- mixture)

CAS: 54410-90-1; MDL No: MFCD01017412; RTECS: GW0680000 Liquid; Linear Formula: CH3(CH2)4C6H10OH; Molecular Formula: C11H22O; MW: 170.29 Density (g/mL): 0.893


  • RTECS: GW0680000
  • Liquid

Formula: C₁₁H₂₂O
MW: 170.30 g/mol
Density: 0.893 g/mL
MDL Number: MFCD01017412
CAS Number: 54410-90-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR