cis-Stilbene ≥97%
Supplier: BeanTown Chemical
Synonyms:
cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene
138935-1G
138935-5G
BT138935-1GEA
21
USD
BT138935-1G
BT138935-5G
cis-Stilbene ≥97%
cis-Stilbene
CAS: 645-49-8; EC No: 211-445-7; MDL No: MFCD00004788; RTECS: DA0513600
Liquid; Molecular Formula: C14H12; MW: 180.25
Boiling Point: 82-84°/0.4 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.011; Refractive Index: 1.622
- RTECS: DA0513600
- Refractive index: 1.622
- Liquid
Formula:
(C₆H₅)CHCH(C₆H₅) MW: 180.25 g/mol Boiling Pt: 82…84 °C/0.4 mmHg Density: 1.011 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00004788 CAS Number: 645-49-8 EINECS: 211-445-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...