4-Bromo-2-ethyl-1-iodobenzene ≥97% stabilized

Supplier: BeanTown Chemical

138880-1G 138880-5G
BT138880-1GEA 27 USD
BT138880-1G BT138880-5G
4-Bromo-2-ethyl-1-iodobenzene ≥97% stabilized
4-Bromo-2-ethyl-1-iodobenzene

CAS: 175278-30-5; MDL No: MFCD00221458 Liquid; Molecular Formula: C8H8BrI; MW: 310.97 Light Sensitive


  • Light Sensitive
  • Liquid

Formula: C₈H₈BrI
MW: 310.97 g/mol
MDL Number: MFCD00221458
CAS Number: 175278-30-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR