Polyethylene glycol 2,000
Supplier: BeanTown Chemical
Synonyms:
PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000
138875-1KG
138875-250G
BT138875-1KGEA
149.97
USD
BT138875-1KG
BT138875-250G
Polyethylene glycol 2,000
Polyethylene glycol 2,000
CAS: 25322-68-3; MDL No: MFCD01779611; RTECS: TQ4041000
Powder; Linear Formula: H(OCH2CH2)nOH; MW ≈2,000
Melting Point: 50-53°; Flash point: 229°C (444°F)
- RTECS: TQ4041000
- Powder
Formula:
H(OCH₂CH₂)nOH MW: 2000 g/mol Melting Pt: 50…53 °C Flash Pt: 229 °C (444 °F) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...