2-Iodoaniline ≥98%

Supplier: BeanTown Chemical

138845-25G 138845-50G 138845-5G
BT138845-25GEA 121 USD
BT138845-25G BT138845-50G BT138845-5G
2-Iodoaniline ≥98%
2-Iodoaniline
Formula: C₆H₆IN
Melting Pt: 55…58 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR