Diethyl diallylmalonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Diethyl 2,2-diallylmalonate, Diallylmalonic Acid Diethyl Ester
138820-10G
138820-250G
138820-50G
BT138820-10GEA
60.46
USD
BT138820-10G
BT138820-250G
BT138820-50G
Diethyl diallylmalonate ≥98%
Diethyl diallylmalonate
CAS: 3195-24-2; EC No: 221-696-4; MDL No: MFCD00009126
Solid; Molecular Formula: C11H20O4; MW: 240.30
Boiling Point: 128-130°/12 mmHg; Flash point:
- Refractive index: 1.446
- Solid
Formula:
C₁₃H₂₀O₄ MW: 240.30 g/mol Boiling Pt: 128…130 °C/12 mmHg Density: 0.994 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00009126 CAS Number: 3195-24-2 EINECS: 221-696-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...