Homophthalic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-(Carboxymethyl)benzoic acid
138810-100G
138810-25G
BT138810-100GEA
224
USD
BT138810-100G
BT138810-25G
Homophthalic acid ≥99%
Homophthalic acid
CAS: 89-51-0; EC No: 201-913-9; MDL No: MFCD00004326; RTECS: CY1575590
Crystalline/Powder; Linear Formula: HOOCCH2C6H4COOH; Molecular Formula: C9H8O4; MW: 180.16
Melting Point: 178-182° (decomposes)
- RTECS: CY1575590
- Crystalline/Powder
Formula:
C₉H₈O₄ Melting Pt: 178…182 °C (dec.) |
MDL Number:
MFCD00004326 CAS Number: 89-51-0 EINECS: 201-913-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...