HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%

Supplier: BeanTown Chemical

Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate

138785-1G 138785-25G 138785-5G
BT138785-1GEA 58.47 USD
BT138785-1G BT138785-25G BT138785-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99%
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

CAS: 148893-10-1; MDL No: MFCD00274639 Crystalline; Molecular Formula: C10H15F6N6OP; MW: 380.23 Melting Point: 183-185° Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₁₀H₁₅F₆N₆OP
Melting Pt: 183…185 °C
MDL Number: MFCD00274639
CAS Number: 148893-10-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR