4-Iodo-m-xylene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,4-Dimethyliodobenzene, 1-Iodo-2,4-dimethylbenzene, 2,4-Dimethyl-1-iodobenzene, 1,3-Dimethyl-4-iodobenzene

138735-10G 138735-50G
BT138735-10GEA 55 USD
BT138735-10G BT138735-50G
4-Iodo-m-xylene ≥98%
4-Iodo-m-xylene

CAS: 4214-28-2; EC No: 224-141-4; MDL No: MFCD00013706 Liquid; Molecular Formula: C8H9I; MW: 232.07 Flash point: 110°C (230°F) Density (g/mL): 1.628; Refractive Index: 1.598 Light Sensitive


  • Refractive index: 1.598
  • Light Sensitive
  • Liquid

Formula: C₈H₉I
MW: 232.06 g/mol
Density: 1.628 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013706
CAS Number: 4214-28-2
EINECS: 224-141-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR