Sodium-4-aminobenzoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
4-Aminobenzoic Acid Sodium Salt, Sodium 4-aminobenzoate
138685-100G
138685-250G
138685-25G
BT138685-100GEA
100
USD
BT138685-100G
BT138685-250G
BT138685-25G
Sodium-4-aminobenzoate ≥99%
Sodium-4-aminobenzoate
CAS: 555-06-6; EC No: 209-080-3; MDL No: MFCD00064395; RTECS: DG2975000
Powder; Molecular Formula: C7H6NNaO2; MW: 159.12
- RTECS: DG2975000
- Powder
Formula:
C₇H₇NO₂ MW: 159.12 g/mol |
MDL Number:
MFCD00064395 CAS Number: 555-06-6 EINECS: 209-080-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...