5-Bromopyridine-2-carbonitrile ≥95%

Supplier: BeanTown Chemical

Synonyms: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

138670-1G 138670-5G
BT138670-1GEA 57 USD
BT138670-1G BT138670-5G
5-Bromopyridine-2-carbonitrile ≥95%
5-Bromopyridine-2-carbonitrile

CAS: 97483-77-7; MDL No: MFCD00234144 Solid; Molecular Formula: C6H3BrN2 ; MW: 183.01 Melting Point: 128-132°; Boiling Point: 100-110°/3 mmHg


  • Solid

Formula: C₆H₃BrN₂
MW: 183.01
Boiling Pt: 100…110 °C/3 mmHg
Melting Pt: 128…132 °C
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR