2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium

138655-100ML 138655-2.5L 138655-500ML
BT138655-100MLEA 52 USD
BT138655-100ML BT138655-2.5L BT138655-500ML
2-Mercaptopyridine-N-oxide sodium salt 40% (w/w) in water
2-Mercaptopyridine N-oxide sodium salt

CAS: 3811-73-2; EC No: 223-296-5; MDL No: MFCD01941547; RTECS: UT9000000 Liquid; Molecular Formula: C5H4NNaOS; MW: 149.15 Density (g/mL): 1.24


  • RTECS: UT9000000
  • Liquid

Formula: C₅H₄NNaOS
MW: 149.15 g/mol
Density: 1.24 g/mL
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR