Titanium(IV) oxide rutile ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile

138645-1KG 138645-250G 138645-5KG
BT138645-1KGEA 79 USD
BT138645-1KG BT138645-250G BT138645-5KG
Titanium(IV) oxide rutile ≥99.5% (trace metals basis)
Titanium(IV) oxide rutile

CAS: 1317-80-2; EC No: 215-282-2; MDL No: MFCD00011269; RTECS: XR2275000 1.0-2.0 Micron APS Powder; Linear Formula: TiO2; MW: 79.90 Melting Point: 1830-1850°; Boiling Point: 2500-3000° Density (g/mL): 4.23


  • RTECS: XR2275000
  • 1.0-2.0 Micron APS Powder

Formula: O₂Ti
MW: 79.88 g/mol
Boiling Pt: 2500…3000 °C
Melting Pt: 1830…1850 °C
Density: 4.23 g/mL
MDL Number: MFCD00011269
CAS Number: 1317-80-2
EINECS: 215-282-2

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR