Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II)

Supplier: BeanTown Chemical
Warning

Synonyms: DIchloro[bis(diphenylphosphinophenyl)ether]palladium(II)

138595-1G 138595-5G
BT138595-1GEA 100 USD
BT138595-1G BT138595-5G
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II)
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II)

CAS: 205319-06-8; MDL No: MFCD09953446 Powder; Molecular Formula: C36H28Cl2OP2Pd; MW: 715.88


  • Powder

Formula: C₃₆H₂₈Cl₂OP₂Pd
MW: 715.887 g/mol
MDL Number: MFCD09953446
CAS Number: 205319-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR