4-Chloro-3-fluorobenzaldehyde ≥98%
Supplier: BeanTown Chemical
138570-1G
138570-25G
138570-5G
BT138570-1GEA
29
USD
BT138570-1G
BT138570-25G
BT138570-5G
4-Chloro-3-fluorobenzaldehyde ≥98%
CAS: 5527-95-7; MDL No: MFCD00143288
Solid; Molecular Formula: C7H4ClFO; MW: 158.56
Melting Point: 46-49°; Flash point: 96°C (205°F)
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₇H₄ClFO Melting Pt: 46…49 °C Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00143288 CAS Number: 5527-95-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...