Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97%

Supplier: BeanTown Chemical

Synonyms: NTCDA, Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 1,4,5,8-Naphthalenetetracarboxylic dianhydride

138535-25G 138535-5G
BT138535-25GEA 156.69 USD
BT138535-25G BT138535-5G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97%
Naphthalene-1,4,5,8-tetracarboxylic dianhydride

CAS: 81-30-1; EC No: 201-342-5; MDL No: MFCD00006915; RTECS: QK3695000 Powder; Molecular Formula: C14H4O6; MW: 268.18 Melting Point: 300° Moisture Sensitive


  • RTECS: QK3695000
  • Moisture Sensitive
  • Powder

Formula: C₁₄H₄O₆
MW: 268.18 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006915
CAS Number: 81-30-1
EINECS: 201-342-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR