Ethyl-6-bromo-4-chloroquinoline-3-carboxylate ≥97%

Supplier: BeanTown Chemical

138485-1G 138485-250MG
BT138485-1GEA 140 USD
BT138485-1G BT138485-250MG
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate ≥97%
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate

CAS: 206257-39-8 Solid; Molecular Formula: C12H9BrClNO2; MW: 314.56 Melting Point: 100-103°


  • Solid

Formula: C₁₂H₉BrClNO₂
Melting Pt: 100…103 °C
MDL Number: MFCD00173367
CAS Number: 206257-39-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR