di-Lithium tetraborate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Lithium tetraborate, Dilithium tetraborate
138455-100G
138455-1KG
138455-25G
BT138455-100GEA
149
USD
BT138455-100G
BT138455-1KG
BT138455-25G
di-Lithium tetraborate ≥99.995% (trace metals basis)
di-Lithium tetraborate
CAS: 12007-60-2; EC No: 234-514-3; MDL No: MFCD00011083
-22 Mesh Powder; Linear Formula: Li2B4O7; MW: 169.12
Melting Point: 760-880°
- -22 Mesh Powder
Formula:
Li₂B₄O₇ MW: 169.12 g/mol Melting Pt: 760…880 °C |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 EINECS: 234-514-3 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...