Lanthanum(III) chloride heptahydrate ≥99%
Supplier: BeanTown Chemical
138405-100G
138405-2.5KG
138405-500G
BT138405-100GEA
64.29
USD
BT138405-100G
BT138405-2.5KG
BT138405-500G
Lanthanum(III) chloride heptahydrate ≥99%
Lanthanum(III) chloride heptahydrate
CAS: 10025-84-0; EC No: 233-237-5; MDL No: MFCD00149756; RTECS: OE4470000
Crystalline; Linear Formula: LaCl3·7H2O; MW: 371.37
Melting Point: 91° (decomposes)
- RTECS: OE4470000
- Crystalline
Formula:
LaCl₃·7H₂O MW: 371.37 g/mol Melting Pt: 96 (dec.) |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...