Tetracycline hydrochloride ≥96%
Supplier: BeanTown Chemical
138385-100G
138385-25G
BT138385-100GEA
150.99
USD
BT138385-100G
BT138385-25G
Tetracycline hydrochloride ≥96%
Tetracycline hydrochloride
CAS: 64-75-5; EC No: 200-593-8; MDL No: MFCD00078142; RTECS: QI9100000
Crystalline/Powder; Molecular Formula: C22H24N2O8·HCl; MW: 480.90
Melting Point: 220-223°
Air Sensitive, Light Sensitive
- RTECS: QI9100000
Formula:
C₂₂H₂₅ClN₂O₈ MW: 480.9 g/mol |
MDL Number:
MFCD00078142 CAS Number: 64-75-5 EINECS: 200-593-8 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...