6-Bromo-1-indanone ≥97%

Supplier: BeanTown Chemical

138380-1G 138380-5G
BT138380-1GEA 55 USD
BT138380-1G BT138380-5G
6-Bromo-1-indanone ≥97%
6-Bromo-1-indanone

  • Solid

Formula: C₉H₇BrO
MW: 211.06 g/mol
Melting Pt: 111…115 °C
MDL Number: MFCD02179286
CAS Number: 14548-39-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR