Zirconium disilicide ≥99.5% (trace metals basis excluding Hf) may contain other phases
Supplier: BeanTown Chemical
138370-100G
138370-25G
BT138370-100GEA
247
USD
BT138370-100G
BT138370-25G
Zirconium disilicide ≥99.5% (trace metals basis excluding Hf) may contain other phases
Zirconium disilicide
CAS: 12039-90-6; EC No: 234-911-1; MDL No: MFCD00151547
-325 Mesh Powder; Linear Formula: ZrSi2; MW: 147.39
Density (g/mL): 4.88
- -325 Mesh Powder
Formula:
Si₂Zr Density: 4.88 g/mL |
MDL Number:
MFCD00151547 CAS Number: 12039-90-6 EINECS: 234-911-1 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...