Sodium iodide ≥99% (dry weight)
Supplier: BeanTown Chemical
CAS: 7681-82-5; EC No: 231-679-3; MDL No: MFCD00003532; RTECS: WB6475000
Crystalline; Linear Formula: NaI; MW: 149.89
Melting Point: 661°; Boiling Point: 1300-1304°
Density (g/mL): 3.67; Refractive Index: 1.775
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: WB6475000
- Refractive index: 1.775
- Air Sensitive, Hygroscopic, Light Sensitive
- Crystalline
Formula:
NaI Boiling Pt: 1300…1304 °C Melting Pt: 661 °C Density: 3.67 g/mL |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...