N-(tert-Butoxycarbonyl)-L-prolinamide ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl (S)-2-Carbamoyl-1-pyrrolidinecarboxylate, (S)-2-Carbamoyl-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, 1-Boc-L-prolinamide, N-Boc-L-prolinamide, Boc-Pro-NH2

138350-1G 138350-250MG 138350-5G
BT138350-1GEA 81 USD
BT138350-1G BT138350-250MG BT138350-5G
N-(tert-Butoxycarbonyl)-L-prolinamide ≥97%
N-(tert-Butoxycarbonyl)-L-prolinamide

CAS: 35150-07-3; EC No: 259-189-5; MDL No: MFCD00190827 Solid; Molecular Formula: C10H18N2O3 ; MW: 214.26 Melting Point: 104-108°


  • Solid

Formula: C₁₀H₁₈N₂O₃
MW: 214.27 g/mol
Melting Pt: 104…108 °C
MDL Number: MFCD00190827
CAS Number: 35150-07-3
EINECS: 259-189-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR