Tetrabutylthiuram disulfide ≥98%

Supplier: BeanTown Chemical

Synonyms: Bis(dibutylthiocarbamoyl) Disulphide

138345-100G 138345-25G
BT138345-100GEA 122 USD
BT138345-100G BT138345-25G
Tetrabutylthiuram disulfide ≥98%
Tetrabutylthiuram disulfide

CAS: 1634-02-2; EC No: 216-652-6; MDL No: MFCD00027191; RTECS: JO0800000 Liquid; Molecular Formula: C18H36N2S4 ; MW: 408.75 Density (g/mL): 1.06 Air Sensitive


  • RTECS: JO0800000
  • Air Sensitive
  • Liquid

Formula: C₁₈H₃₆N₂S₄
MW: 408.74 g/mol
Density: 1.06 g/mL
MDL Number: MFCD00027191
CAS Number: 1634-02-2
EINECS: 216-652-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR