Sodium hexachlororhodate (III) dodecahydrate
Supplier: BeanTown Chemical
Synonyms:
Rhodium sodium chloride dodecahydrate
138330-1G
138330-5G
BT138330-1GEA
204
USD
BT138330-1G
BT138330-5G
Sodium hexachlororhodate (III) dodecahydrate
Sodium hexachlororhodate (III) dodecahydrate
CAS: 14972-70-4; EC No: 239-047-9; MDL No: MFCD00003509
Crystalline; Linear Formula: Na3RhCl6·12H2O; MW: 600.77
Melting Point: 900° (decomposes)
- Crystalline
Formula:
Cl₆Na₃Rh·12H₂O MW: 600.77 g/mol Melting Pt: 900 °C (dec.) |
MDL Number:
MFCD00003509 CAS Number: 14972-70-4 EINECS: 239-047-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...