Cyclopropanecarboxamide ≥98%

Supplier: BeanTown Chemical

138325-25G 138325-5G
BT138325-25GEA 99 USD
BT138325-25G BT138325-5G
Cyclopropanecarboxamide ≥98%
Cyclopropanecarboxamide

CAS: 6228-73-5; EC No: 228-332-3; MDL No: MFCD00013729 Crystalline/Powder; Molecular Formula: C4H7NO; MW: 85.11 Melting Point: 122-126°


  • Crystalline/Powder

Formula: C₄H₇NO
MW: 85.11 g/mol
Melting Pt: 122…126 °C
MDL Number: MFCD00013729
CAS Number: 6228-73-5
EINECS: 228-332-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR