Biphenyl-3-carboxaldehyde ≥95%
Supplier: BeanTown Chemical
Synonyms:
3-Phenylbenzaldehyde, [1,1'-Biphenyl]-3-carbaldehyde
138310-1G
138310-250MG
BT138310-1GEA
169
USD
BT138310-1G
BT138310-250MG
Biphenyl-3-carboxaldehyde ≥95%
Biphenyl-3-carboxaldehyde
CAS: 1204-60-0; MDL No: MFCD01740432; RTECS: DV1771500
Liquid; Molecular Formula: C13H10O; MW: 182.22
Density (g/mL): 1.118
Air Sensitive
- RTECS: DV1771500
- Air Sensitive
- Liquid
Formula:
C₁₃H₁₀O Density: 1.118 g/mL |
MDL Number:
MFCD01740432 CAS Number: 1204-60-0 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...