Tricine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

138245-100G 138245-25G 138245-500G
BT138245-100GEA 75.87 USD
BT138245-100G BT138245-25G BT138245-500G
Tricine ≥98%
Tricine

CAS: 5704-04-1; EC No: 227-193-6; MDL No: MFCD00004277 Crystalline; Linear Formula: (HOCH2)3CNHCH2COOH; Molecular Formula: C6H13NO5; MW: 179.17 Melting Point: 187° (decomposes)


  • Crystalline

Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 187 °C (dec.)
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR