Ammonium formate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Formic acid ammonium salt
138235-250G
138235-50G
BT138235-250GEA
59.58
USD
BT138235-250G
BT138235-50G
Ammonium formate ≥99%
Ammonium formate
CAS: 540-69-2; EC No: 208-753-9; MDL No: MFCD00013103; RTECS: BQ6650000
Crystalline; Linear Formula: HCO2NH4; MW: 63.06
Melting Point: 119-121°
Density (g/mL): 1.28
Hygroscopic
- RTECS: BQ6650000
- Hygroscopic
- Crystalline
Formula:
HCO₂NH₄ MW: 63.06 g/mol Melting Pt: 119…121 °C Density: 1.28 g/mL |
MDL Number:
MFCD00013103 CAS Number: 540-69-2 EINECS: 208-753-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...