Pig Hemin
Supplier: BeanTown Chemical
138155-1G
138155-25G
138155-5G
BT138155-1GEA
45.01
USD
BT138155-1G
BT138155-25G
BT138155-5G
Pig Hemin
Proteins and Peptides
EC No: 240-140-1
RTECS: LJ8080000
Molecular formula: C₃₄H₃₂ClFeN₄O₄
MW: 651.95
Merck Number: 14,4644
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...