2-Methoxy-4-vinylphenol ≥97% stabilized
Supplier: BeanTown Chemical
138140-1G
138140-5G
BT138140-1GEA
106
USD
BT138140-1G
BT138140-5G
2-Methoxy-4-vinylphenol ≥97% stabilized
2-Methoxy-4-vinylphenol
CAS: 7786-61-0; EC No: 232-101-2; MDL No: MFCD00015437; RTECS: SL8205000
Liquid; Molecular Formula: C9H10O2; MW: 150.17
Boiling Point: 224°
Density (g/mL): 1.11; Refractive Index: 1.582
- RTECS: SL8205000
- Refractive index: 1.582
- Liquid
Formula:
C₉H₁₀O₂ Boiling Pt: 224 °C Density: 1.11 g/mL |
MDL Number:
MFCD00015437 CAS Number: 7786-61-0 EINECS: 232-101-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...