Perylene ≥98%
Supplier: BeanTown Chemical
138135-1G
138135-5G
BT138135-1GEA
35.17
USD
BT138135-1G
BT138135-5G
Perylene ≥98%
Perylene
CAS: 198-55-0; EC No: 205-900-9; MDL No: MFCD00004142; RTECS: SE3794000
Crystalline; Molecular Formula: C20H12; MW: 252.32
Melting Point: 276-280°
- RTECS: SE3794000
- Crystalline
Formula:
C₂₀H₁₂ MW: 252.32 g/mol Melting Pt: 276…280 °C |
MDL Number:
MFCD00004142 CAS Number: 198-55-0 EINECS: 205-900-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...