2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid

138130-100G 138130-25G 138130-500G
BT138130-100GEA 89.01 USD
BT138130-100G BT138130-25G BT138130-500G
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid ≥99%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

CAS: 10191-18-1; EC No: 233-465-5; MDL No: MFCD00007533; RTECS: KI7363500 Crystalline; Molecular Formula: C6H15NO5S; MW: 213.25


  • RTECS: KI7363500
  • Crystalline

Formula: C₆H₁₅NO₅S
MW: 213.25 g/mol
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR