Copper(II) trifluoroacetate hydrate

Supplier: BeanTown Chemical
Warning

Synonyms: Copper(II) 2,2,2-trifluoroacetate hydrate

138120-10G 138120-50G
BT138120-10GEA 57 USD
BT138120-10G BT138120-50G
Copper(II) trifluoroacetate hydrate
Copper(II) trifluoroacetate hydrate

CAS: 123333-88-0; MDL No: MFCD00013200 Crystalline; Linear Formula: Cu(OOCCF3)2·xH2O; MW: 289.57 (anhydrous)


  • Crystalline

Formula: C₄CuF₆O₄·xH₂O
MW: 289.576 g/mol (anhy)
MDL Number: MFCD00013200
CAS Number: 123333-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR