5-Methoxysalicylaldehyde ≥98%

Supplier: BeanTown Chemical

138115-25G 138115-50G 138115-5G
BT138115-25GEA 196 USD
BT138115-25G BT138115-50G BT138115-5G
5-Methoxysalicylaldehyde ≥98%
5-Methoxysalicylaldehyde

CAS: 672-13-9; EC No: 211-589-0; MDL No: MFCD00003332; RTECS: BZ2800000 Liquid; Molecular Formula: C8H8O3; MW: 152.15 Melting Point: 4°; Boiling Point: 103°/2.5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.219; Refractive Index: 1.578 Air Sensitive


  • RTECS: BZ2800000
  • Refractive index: 1.578
  • Air Sensitive
  • Liquid

Formula: C₈H₈O₃
Boiling Pt: 103 °C/2.5 mmHg
Melting Pt: 4 °C
Density: 1.219 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR