Methyl cellulose, 8000 cP (2% H₂O)

Supplier: BeanTown Chemical

Synonyms: Cellulose methyl ether

138090-100G 138090-500G
BT138090-100GEA 50 USD
BT138090-100G BT138090-500G
Methyl cellulose, 8000 cP (2% H₂O)
Methyl cellulose

CAS: 9004-67-5; MDL No: MFCD00081763 Powder


  • Powder

MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR