Cinnamyl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

138085-1KG 138085-250G
BT138085-1KGEA 113 USD
BT138085-1KG BT138085-250G
Cinnamyl alcohol ≥98%
Cinnamyl alcohol

CAS: 104-54-1; EC No: 203-212-3; MDL No: MFCD00002921; RTECS: GE2200000 Solid; Linear Formula: C6H5CH=CHCH2OH; Molecular Formula: C9H8O; MW: 134.18 Melting Point: 30-33°; Boiling Point: 250°; Flash point: 126°C (259°F) Density (g/mL): 1.044


  • RTECS: GE2200000
  • Solid

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 250 °C
Melting Pt: 30…33 °C
Density: 1.044 g/mL
Flash Pt: 126 °C (259 °F)
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR