5-(Difluoromethoxy)-2-mercaptobenzimidazole ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-(Difluoromethoxy)-2-benzimidazolethiol, 5-Difluoromethoxy-2-mercaptobenzimidazole, 5-(Difluoromethoxy)-1H-benzo[d]imidazole-2-thiol, 5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

138065-1G 138065-5G
BT138065-1GEA 24 USD
BT138065-1G BT138065-5G
5-(Difluoromethoxy)-2-mercaptobenzimidazole ≥97%
5-(Difluoromethoxy)-2-mercaptobenzimidazole

CAS: 97963-62-7; MDL No: MFCD00467504 Powder; Molecular Formula: C8H6F2N2OS; MW: 216.21 Melting Point: 239-243°


  • Powder

Formula: C₈H₆F₂N₂OS
MW: 216.21 g/mol
Melting Pt: 239…243 °C
MDL Number: MFCD00467504
CAS Number: 97963-62-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR