(±)-Boc-α-phosphonoglycine trimethyl ester ≥95%
Supplier: BeanTown Chemical
138060-1G
138060-25G
138060-5G
BT138060-1GEA
65
USD
BT138060-1G
BT138060-25G
BT138060-5G
(±)-Boc-α-phosphonoglycine trimethyl ester ≥95%
(±)-Boc-α-phosphonoglycine trimethyl ester
CAS: 89524-98-1; MDL No: MFCD02178893
Powder; Molecular Formula: C10H20NO7P; MW: 297.24
Melting Point: 67-71°
- Powder
Formula:
C₁₀H₂₀NO₇P Melting Pt: 67…71 °C |
MDL Number:
MFCD02178893 CAS Number: 89524-98-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...