5-Bromo-2-hydroxypyrimidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromopyrimidin-2(1H)-one, 5-Bromo-2-pyrimidinone

138050-1G 138050-5G
BT138050-1GEA 71 USD
BT138050-1G BT138050-5G
5-Bromo-2-hydroxypyrimidine ≥98%
5-Bromo-2-hydroxypyrimidine

CAS: 38353-06-9; EC No: 253-896-2; MDL No: MFCD00233952 Solid; Molecular Formula: C4H3BrN2O; MW: 174.98 Melting Point: 230° (decomposes)


  • Solid

Formula: C₄H₃BrN₂O
MW: 174.98 g/mol
Melting Pt: 230 °C (dec.)
MDL Number: MFCD01318956
CAS Number: 38353-06-9
EINECS: 253-896-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR