2,2'-Bicinchoninic acid disodium salt
Supplier: BeanTown Chemical
|
Warning
|
138045-1G
138045-5G
BT138045-1GEA
37
USD
BT138045-1G
BT138045-5G
2,2'-Bicinchoninic acid disodium salt
2,2'-Bicinchoninic acid disodium salt
CAS: 979-88-4; MDL No: MFCD00037500
Powder/Chunks; Molecular Formula: C20H10N2Na2O4; MW: 388.28
- Powder/Chunks
Formula:
C₂₀H₁₀N₂Na₂O₄ |
MDL Number:
MFCD00037500 CAS Number: 979-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...