Ethyl benzoylacetate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 3-oxo-3-phenylpropanoate
138035-1KG
138035-250G
138035-50G
BT138035-1KGEA
247
USD
BT138035-1KG
BT138035-250G
BT138035-50G
Ethyl benzoylacetate ≥96%
Ethyl benzoylacetate
CAS: 94-02-0; EC No: 202-295-3; MDL No: MFCD00009196; RTECS: AF4878000
Liquid; Molecular Formula: C11H12O3; MW: 192.21
Boiling Point: 265-270°
Density (g/mL): 1.11; Refractive Index: 1.529
Light Sensitive
- RTECS: AF4878000
- Refractive index: 1.529
- Light Sensitive
- Liquid
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 265 Density: 1.11 g/mL Flash Pt: 140 |
MDL Number:
MFCD00009196 CAS Number: 94-02-0 EINECS: 202-295-3 Merck Index: 13,03803 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...