Tungsten, gauze woven from wire 100 mesh Ø 0.025 mm

Supplier: BeanTown Chemical

138005-150X150 138005-300X300 138005-75X75MM
BT138005-150X1EA 522 USD
BT138005-150X1 BT138005-300X3 BT138005-75X75
Tungsten, gauze woven from wire 100 mesh Ø 0.025 mm
Tungsten

CAS: 7440-33-7; EC No: 231-143-9; MDL No: MFCD00011461 Wire Cloth; Molecular Formula: W; MW: 183.84 Melting Point: 3410°; Boiling Point: 5660° Density (g/mL): 19.3


  • Wire Cloth

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19.3 g/mL
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR