Ninhydrin ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
137965-10G
137965-50G
BT137965-10GEA
23.26
USD
BT137965-10G
BT137965-50G
Ninhydrin ≥99%
Ninhydrin
CAS: 485-47-2; EC No: 207-618-1; MDL No: MFCD00003791; RTECS: NK5425000
Crystalline; Molecular Formula: C9H6O4; MW: 178.14
Melting Point: 250° (decomposes)
Light Sensitive
- RTECS: NK5425000
- Light Sensitive
- Crystalline
Formula:
C₉H₆O₄ Melting Pt: 250 °C (dec.) |
MDL Number:
MFCD00003791 CAS Number: 485-47-2 EINECS: 207-618-1 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...