EDTA dipotassium salt dihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt
CAS: 25102-12-9; EC No: 217-895-0; MDL No: MFCD00150036
Crystalline/Powder; Molecular Formula: C10H14K2N2O8·2H2O; MW: 404.46
Melting Point: 255-280° (decomposes)
- Crystalline/Powder
Formula:
C₁₀H₁₄K₂N₂O₈·2H₂O MW: 404.46 g/mol Melting Pt: 272 (dec.) |
MDL Number:
MFCD00150036 CAS Number: 25102-12-9 EINECS: 217-895-0 Merck Index: 13,03546 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...